DL-Homoserine ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A0319-1G
A0319-5G
TCIAA0319-1GEA
82.5
EUR
TCIAA0319-1G
TCIAA0319-5G
DL-Homoserine ≥98.0% (by titrimetric analysis)
DL-Homoserine
Formule:
C₄H₉NO₃ Kookpunt: 276 °C (1013 hPa) Smeltpunt: 151 °C |
MDL Nummer:
MFCD00002618 CAS nummer: 1927-25-9 EINECS: 217-661-8 Merck Index: 14,04739 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...