trans-4-Hydroxy-L(-)-proline ≥99.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH

H0296-25G H0296-5G H0296-100G
TCIAH0296-25GEA 41.8 EUR
TCIAH0296-25G TCIAH0296-5G TCIAH0296-100G
trans-4-Hydroxy-L(-)-proline ≥99.0% (by HPLC, titration analysis)
trans-4-Hydroxy-L(-)-proline
Formule: C₅H₉NO₃
Kookpunt: 296 °C (1013 hPa)
Smeltpunt: 206 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064320
CAS nummer: 51-35-4
EINECS: 200-091-9
Merck Index: 14,04840
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6641c01b551f7d25e459a1ed979f5da62b0833bf&vid=6641c01b551f7d25e459a1ed979f5da62b0833bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystaline
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D -74.0 to -77.0 deg (C=4, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR