2,4-Dichloorfenoxyazijnzuur ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2,4-D

D0396-25G D0396-500G
TCIAD0396-25GEA 30.8 EUR
TCIAD0396-25G TCIAD0396-500G
2,4-Dichloorfenoxyazijnzuur ≥97.0% (by titrimetric analysis)
2,4-Dichloorfenoxyazijnzuur
Formule: (C₆H₃Cl₂)OCH₂COOH
Kookpunt: 346 °C (1013 hPa)
Smeltpunt: 137…139 °C
Dichtheid: 1,563 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004300
CAS nummer: 94-75-7
EINECS: 202-361-1
UN: 3077
ADR: 9,III
Merck Index: 14,02796
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=662c3c76fa537c4599fdbc9eb32064fdb48a8183&vid=662c3c76fa537c4599fdbc9eb32064fdb48a8183&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 138.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR