Ethyl-O-tolylacetate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T1225-25G
T1225-5G
TCIAT1225-25GEA
89.1
EUR
TCIAT1225-25G
TCIAT1225-5G
Ethyl-O-tolylacetate ≥97.0% (by GC)
Ethyl-O-tolylacetate
Formule:
(C₆H₄CH₃)CH₂COOCH₂CH₃ Kookpunt: 77°C Dichtheid: 0,9988 g/cm³ (25 °C) Vlampunt: 107°C |
MDL Nummer:
MFCD00009179 CAS nummer: 40291-39-2 EINECS: 254-871-9 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...