4-CPA ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-CPA

C0250-25G C0250-500G
TCIAC0250-25GEA 17.6 EUR
TCIAC0250-25G TCIAC0250-500G
4-CPA ≥98.0% (by HPLC, titration analysis)
4-CPA
Formule: (C₆H₄Cl)OCH₂COOH
Kookpunt: 308 °C (1013 hPa)
Smeltpunt: 160 °C
Dichtheid: 1,53 g/cm³ (20 °C)
MDL Nummer: MFCD00004305
CAS nummer: 122-88-3
EINECS: 204-581-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c0f52ffcf3a490e71a05e385028422be1b64d89&vid=0c0f52ffcf3a490e71a05e385028422be1b64d89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 157.0 to 161.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR