(Chloromethyl)trimethylsilane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0862-25ML C0862-250ML C0862-100ML
TCIAC0862-25MLEA 35.2 EUR
TCIAC0862-25ML TCIAC0862-250ML TCIAC0862-100ML
(Chloromethyl)trimethylsilane ≥98.0% (by GC)
(Chloromethyl)trimethylsilane
Formule: ClH₂CSi(CH₃)₃
Kookpunt: 98 °C (1013 hPa)
Dichtheid: 0,8861 g/cm³ (20 °C)
Vlampunt: -2°C
MDL Nummer: MFCD00000878
CAS nummer: 2344-80-1
EINECS: 219-058-5
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea4fc314831c53b50d88e5cee5778dd531b2b817&vid=ea4fc314831c53b50d88e5cee5778dd531b2b817&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR