Vinyltrimethylsilane ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

V0067-100ML V0067-25ML
TCIAV0067-100MLEA 263 EUR
TCIAV0067-100ML TCIAV0067-25ML
Vinyltrimethylsilane ≥97.0% (by GC)
Vinyltrimethylsilane
Formule: C₅H₁₂Si
Kookpunt: 56°C
Dichtheid: 0,7038 g/cm³ (20 °C)
Vlampunt: -24°C
MDL Nummer: MFCD00008606
CAS nummer: 754-05-2
EINECS: 212-042-9
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=987618ad45b6bd7d71c074ca9ab68d4c7244d7db&vid=987618ad45b6bd7d71c074ca9ab68d4c7244d7db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR