Benzyl mercaptan ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Toluene-α-thiol, α-Toluenethiol
T0287-25G T0287-500G T0287-100G
TCIAT0287-25GEA 28.6 EUR
TCIAT0287-25G TCIAT0287-500G TCIAT0287-100G
Benzyl mercaptan ≥96.0% (by GC)
Benzyl mercaptan
Formule: C₇H₈S
Kookpunt: 195°C
Smeltpunt: 29 °C
Dichtheid: 1,055 g/cm³ (20 °C)
Vlampunt: 70°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004867
CAS nummer: 100-53-8
EINECS: 202-862-5
UN: 2810
ADR: 6.1,III
Merck Index: 14,09322
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0531666c553a72328df391f30fa66bc82f276fc&vid=c0531666c553a72328df391f30fa66bc82f276fc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %