Benzyltributylammonium chloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N-Benzyl-N,N-dibutylbutan-1-aminium chloride
B1384-25G
B1384-500G
TCIAB1384-25GEA
17.6
EUR
TCIAB1384-25G
TCIAB1384-500G
Benzyltributylammonium chloride ≥98.0% (by titrimetric analysis)
Benzyltributylammonium chloride
Formule:
C₁₉H₃₄ClN Smeltpunt: 155…159 °C |
MDL Nummer:
MFCD00011849 CAS nummer: 23616-79-7 EINECS: 245-787-3 UN: 1759 ADR: 8,III |
|
Specification Test Results
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...