2-(Trifluoromethyl)benzylamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: (2-(Trifluoromethyl)phenyl)methanamine
T2315-25G T2315-5G
TCIAT2315-25GEA 208 EUR
TCIAT2315-25G TCIAT2315-5G
2-(Trifluoromethyl)benzylamine ≥98.0% (by GC, titration analysis)
2-(Trifluoromethyl)benzylamine
Formule: C₈H₈F₃N
MW: 175.15 g/mol
Kookpunt: 110°C
Vlampunt: 69°C
MDL Nummer: MFCD00010297
CAS nummer: 3048-01-9
EINECS: 221-258-2
UN: 2735
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70a64351b8cbaae8de2e074332d973e96ac14578&vid=70a64351b8cbaae8de2e074332d973e96ac14578&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %