ω-Chloro-2,4-difluoroacetophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone

C2030-25G C2030-5G
TCIAC2030-25GEA 95.7 EUR
TCIAC2030-25G TCIAC2030-5G
ω-Chloro-2,4-difluoroacetophenone ≥98.0% (by GC)
ω-Chloro-2,4-difluoroacetophenone
Formule: C₈H₅ClF₂O
MW: 190.58 g/mol
Kookpunt: 233
MDL Nummer: MFCD00013252
CAS nummer: 51336-94-8
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05d0cdccdb3cf0cb1763d30fa57ca5d5bcb029b0&vid=05d0cdccdb3cf0cb1763d30fa57ca5d5bcb029b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR