(4-Methoxyphenyl)acetonitrile ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M0856-25G M0856-500G
TCIAM0856-25GEA 22 EUR
TCIAM0856-25G TCIAM0856-500G
(4-Methoxyphenyl)acetonitrile ≥97.0% (by GC)
(4-Methoxyphenyl)acetonitrile
Formule: C₉H₉NO
Kookpunt: 290°C
Smeltpunt: 8 °C
Dichtheid: 1,085 g/cm³ (20 °C)
Vlampunt: 117°C
MDL Nummer: MFCD00001919
CAS nummer: 104-47-2
EINECS: 203-206-0
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db7859754b948a613db6fc70e43deff8bd1a28ec&vid=db7859754b948a613db6fc70e43deff8bd1a28ec&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %