3-Fluoropyrocatechol ≥98.0% (by GC), Technical Grade
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
F0282-1G F0282-5G
TCIAF0282-1GEA 29.7 EUR
TCIAF0282-1G TCIAF0282-5G
3-Fluoropyrocatechol ≥98.0% (by GC), Technical Grade
3-Fluoropyrocatechol
Formule: C₆H₅FO₂
Kookpunt: 218 °C (1013 hPa)
Smeltpunt: 71 °C
Dichtheid: 1,514 g/cm³ (20 °C)
MDL Nummer: MFCD00042582
CAS nummer: 363-52-0
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a621a5cacaff2c8fa099c7c240dec318a8edf06&vid=ab58614c4a0d69f305659b1e898d3ed70b87df20&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C