2,5-Dimethyl-p-phenylenediamine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2183-25G D2183-100G
TCIAD2183-25GEA 69.3 EUR
TCIAD2183-25G TCIAD2183-100G
2,5-Dimethyl-p-phenylenediamine ≥98.0% (by GC, titration analysis)
2,5-Dimethyl-p-phenylenediamine
Formule: C₈H₁₂N₂
MDL Nummer: MFCD00052675
CAS nummer: 6393-01-7
EINECS: 229-003-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b803b488a37dd1cb208d53b3761a2d5894d3a70&vid=5b803b488a37dd1cb208d53b3761a2d5894d3a70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR