1-Amino-2-methylbutane ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-Methylbutylamine, 2-Methyl-1-butylamine
M1263-25ML M1263-5ML
TCIAM1263-25MLEA 204 EUR
TCIAM1263-25ML TCIAM1263-5ML
1-Amino-2-methylbutane ≥98.0% (by GC)
1-Amino-2-methylbutane
Formule: C₅H₁₃N
MW: 87.16 g/mol
Kookpunt: 97°C
Vlampunt: 3°C
MDL Nummer: MFCD00008147
CAS nummer: 96-15-1
EINECS: 202-483-5
UN: 2733
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac12c8247d3a9ddd34d16f297b664970e566e04e&vid=ac12c8247d3a9ddd34d16f297b664970e566e04e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %