4-Chloro-m-anisidine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C1774-1G C1774-5G
TCIAC1774-1GEA 24.2 EUR
TCIAC1774-1G TCIAC1774-5G
4-Chloro-m-anisidine ≥98.0% (by GC, titration analysis)
4-Chloro-m-anisidine
Formule: C₇H₈ClNO
MDL Nummer: MFCD00672967
CAS nummer: 13726-14-2
UN: 2233
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0279376c19264565cbd5ba81f7f10ac9676e1ba0&vid=0279376c19264565cbd5ba81f7f10ac9676e1ba0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR