5-Bromovaleronitrile ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B1453-25G
B1453-5G
TCIAB1453-25GEA
126
EUR
TCIAB1453-25G
TCIAB1453-5G
5-Bromovaleronitrile ≥97.0% (by GC)
5-Bromovaleronitrile
Formule:
BrH₂C(CH₂)₂CH₂CN Kookpunt: 111°C Dichtheid: 1,3775 g/cm³ (20 °C) |
MDL Nummer:
MFCD00001976 CAS nummer: 5414-21-1 EINECS: 226-505-8 UN: 3276 ADR: 6.1,III |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...