3-Chloro-4-fluorobenzotrifluoride ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C1515-25G C1515-5G
TCIAC1515-25GEA 185 EUR
TCIAC1515-25G TCIAC1515-5G
3-Chloro-4-fluorobenzotrifluoride ≥98.0% (by GC)
3-Chloro-4-fluorobenzotrifluoride
Formule: C₇H₃ClF₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00042207
CAS nummer: 78068-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e61bfa960a93aca75bfa913bd49d406834dfe07&vid=8e61bfa960a93aca75bfa913bd49d406834dfe07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR