o-Toluonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Methylbenzonitrile, o-Methylbenzonitrile, o-Tolunitrile

T0306-25G T0306-500G
TCIAT0306-25GEA 17.6 EUR
TCIAT0306-25G TCIAT0306-500G
o-Toluonitrile ≥98.0% (by GC)
o-Toluonitrile
Formule: C₈H₇N
Kookpunt: 205°C
Smeltpunt: –13 °C
Dichtheid: 0,99 g/cm³ (20 °C)
Vlampunt: 82°C
MDL Nummer: MFCD00001792
CAS nummer: 529-19-1
EINECS: 208-451-7
Merck Index: 14,09537
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42acc0edd599cf025c970d4d6c15f080876c3715&vid=42acc0edd599cf025c970d4d6c15f080876c3715&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR