4,4'-Difluorobenzophenone ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Bis(4-Fluorophenyl)methanone, Bis(4-fluorophenyl) ketone
D1797-25G D1797-500G D1797-100G
TCIAD1797-25GEA 22 EUR
TCIAD1797-25G TCIAD1797-500G TCIAD1797-100G
4,4'-Difluorobenzophenone ≥99.0% (by GC)
4,4'-Difluorobenzophenone
Formule: C₁₃H₈F₂O
MW: 218.2 g/mol
MDL Nummer: MFCD00000353
CAS nummer: 345-92-6
EINECS: 206-466-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a289d5230f554dfb16215bf76480614f073d2af&vid=7a289d5230f554dfb16215bf76480614f073d2af&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 107.0 to 110.0 deg-C
Solubility in Toluene almost transparency