1-Methylbenzimidazole ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-Methyl-1H-benzimidazole

M2173-25G M2173-5G
TCIAM2173-25GEA 208 EUR
TCIAM2173-25G TCIAM2173-5G
1-Methylbenzimidazole ≥98.0% (by GC, titration analysis)
1-Methylbenzimidazole
Formule: C₈H₈N₂
Kookpunt: 154°C
Smeltpunt: 59…62 °C
Dichtheid: 1,11 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00192275
CAS nummer: 1632-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b86b75a6043d69690dd3dabf507a5f226d51cde&vid=1b86b75a6043d69690dd3dabf507a5f226d51cde&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 58.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR