2-Thiophenemethanol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Thiophen-2-methanol, Thiophen-2-ylmethanol, 2-Thienylmethanol
T1112-25G T1112-500G T1112-100G
TCIAT1112-25GEA 92.4 EUR
TCIAT1112-25G TCIAT1112-500G TCIAT1112-100G
2-Thiophenemethanol ≥98.0% (by GC)
2-Thiophenemethanol
Formule: C₅H₆OS
Kookpunt: 207°C
Smeltpunt: 0 °C
Dichtheid: 1,206 g/cm³ (20 °C)
MDL Nummer: MFCD00005454
CAS nummer: 636-72-6
EINECS: 211-264-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ada7e7146bc0e468ee46093a0ecda1c6a7255a7&vid=1ada7e7146bc0e468ee46093a0ecda1c6a7255a7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %