2,4,5-Trimethylthiazole ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T1068-25G T1068-5G
TCIAT1068-25GEA 72.6 EUR
TCIAT1068-25G TCIAT1068-5G
2,4,5-Trimethylthiazole ≥97.0% (by GC, titration analysis)
2,4,5-Trimethylthiazole
Formule: C₆H₉NS
Kookpunt: 165 °C (1013 hPa)
Dichtheid: 1,013 g/cm³ (16 °C)
Vlampunt: 56°C
MDL Nummer: MFCD00005332
CAS nummer: 13623-11-5
EINECS: 237-107-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=040ea2f0499a4b1e7016484273262a7daf186cdc&vid=040ea2f0499a4b1e7016484273262a7daf186cdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR