5-Nitrouracil ≥99.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Nitrouracil, 5-Nitropyrimidine-2,4(1H,3H)-dione, 2,4-Dihydroxy-5-nitropyrimidine

N0281-25G N0281-5G
TCIAN0281-25GEA 37.4 EUR
TCIAN0281-25G TCIAN0281-5G
5-Nitrouracil ≥99.0% (by HPLC, titration analysis)
5-Nitrouracil
Formule: C₄H₃N₃O₄
MW: 157.09 g/mol
Smeltpunt: 285°C
MDL Nummer: MFCD00006021
CAS nummer: 611-08-5
EINECS: 210-250-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0642a5bd0622bc1582cb0ce9eab4768be640615f&vid=0642a5bd0622bc1582cb0ce9eab4768be640615f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR