2,5-Dimethyl-3-hexyne-2,5-diol ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D0738-25G
D0738-500G
TCIAD0738-25GEA
17.6
EUR
TCIAD0738-25G
TCIAD0738-500G
2,5-Dimethyl-3-hexyne-2,5-diol ≥99.0% (by GC)
2,5-Dimethyl-3-hexyne-2,5-diol
Formule:
C₈H₁₄O₂ Kookpunt: 205 °C (1013 hPa) Smeltpunt: 93…95 °C Dichtheid: 0,9469 g/cm³ (20 °C) |
MDL Nummer:
MFCD00004468 CAS nummer: 142-30-3 EINECS: 205-533-4 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 93.0 to 96.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...