2-Amino-4-methylthiazool ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A1303-25G A1303-250G
TCIAA1303-25GEA 28.6 EUR
TCIAA1303-25G TCIAA1303-250G
2-Amino-4-methylthiazool ≥99.0% (by GC)
2-Amino-4-methylthiazool
Formule: C₄H₆N₂S
Kookpunt: 232°C
Smeltpunt: 42 °C
Dichtheid: 1,202 g/cm³ (50 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005329
CAS nummer: 1603-91-4
EINECS: 216-505-6
Merck Index: 14,00450
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecf83f2ee4c1026820fbccb2391e8b72e5f55c0f&vid=ecf83f2ee4c1026820fbccb2391e8b72e5f55c0f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 43.0 to 46.0 deg-C