6-Chloronicotinonitrile ≥97.0% (by GC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Chloro-5-cyanopyridine, 6-Chloro-3-pyridinecarbonitrile, 2-Chloropyridine-5-carbonitrile
C2056-25G
C2056-5G
TCIAC2056-25GEA
174
EUR
TCIAC2056-25G
TCIAC2056-5G
6-Chloronicotinonitrile ≥97.0% (by GC, total nitrogen)
6-Chloronicotinonitrile
Formule:
C₆H₃ClN₂ Kookpunt: 107°C |
MDL Nummer:
MFCD00084941 CAS nummer: 33252-28-7 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...