2-Chlorobenzothiazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0332-25G C0332-250G
TCIAC0332-25GEA 39.6 EUR
TCIAC0332-25G TCIAC0332-250G
2-Chlorobenzothiazole ≥98.0% (by GC)
2-Chlorbenzothiazol
Formule: C₇H₄ClNS
Kookpunt: 248°C
Smeltpunt: 21…24 °C
Dichtheid: 1,37 g/cm³ (20 °C)
MDL Nummer: MFCD00005776
CAS nummer: 615-20-3
EINECS: 210-415-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4777d13ca7734029d7673aea1d4619c91266238&vid=a4777d13ca7734029d7673aea1d4619c91266238&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR