2-Fluoro-5-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F0289-1G F0289-25G F0289-5G
TCIAF0289-1GEA 45.1 EUR
TCIAF0289-1G TCIAF0289-25G TCIAF0289-5G
2-Fluoro-5-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
2-Fluoro-5-(trifluoromethyl)benzaldehyde
Formule: C₈H₄F₄O
MDL Nummer: MFCD00061311
CAS nummer: 146137-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dd52268a4a22942d8287927053713afd49a05c8&vid=2dd52268a4a22942d8287927053713afd49a05c8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %