2-Propoxybenzoic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-n-Propoxybenzoic acid

P1694-25G P1694-5G
TCIAP1694-25GEA 117 EUR
TCIAP1694-25G TCIAP1694-5G
2-Propoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Propoxybenzoic acid
Formule: C₁₀H₁₂O₃
Kookpunt: 207°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01075687
CAS nummer: 2100-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ff2611f575a1ec527ff489c9a7dc91bcda3e76f&vid=4ff2611f575a1ec527ff489c9a7dc91bcda3e76f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR