3,5-Bis(trifluoromethyl)benzaldehyde ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1751-25G B1751-5G
TCIAB1751-25GEA 173 EUR
TCIAB1751-25G TCIAB1751-5G
3,5-Bis(trifluoromethyl)benzaldehyde ≥95.0% (by GC)
3,5-Bis(trifluoromethyl)benzaldehyde
Formule: C₉H₄F₆O
Kookpunt: 171°C
Smeltpunt: –2 °C
Dichtheid: 1,46 g/cm³ (20 °C)
Vlampunt: 70°C
MDL Nummer: MFCD00010206
CAS nummer: 401-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e50572b568b8a9439d8f5eedf9df7b68ec7e5ebf&vid=e50572b568b8a9439d8f5eedf9df7b68ec7e5ebf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %