Ethyl isovalerate ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0194-25ML I0194-500ML
TCIAI0194-25MLEA 18.7 EUR
TCIAI0194-25ML TCIAI0194-500ML
Ethyl isovalerate ≥99.0% (by GC)
Ethyl isovalerate
Formule: C₇H₁₄O₂
Kookpunt: 134°C
Smeltpunt: –99 °C
Dichtheid: 0,86 g/cm³ (20 °C)
Vlampunt: 31°C
MDL Nummer: MFCD00009203
CAS nummer: 108-64-5
EINECS: 203-602-3
UN: 3272
ADR: 3,III
Merck Index: 14,03816
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7012c7c8b2680e0b8ab04b838e85ec4915829a01&vid=7012c7c8b2680e0b8ab04b838e85ec4915829a01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR