Thiophene-3-carbaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde
T0864-25G
T0864-5G
TCIAT0864-25GEA
168
EUR
TCIAT0864-25G
TCIAT0864-5G
Thiophene-3-carbaldehyde ≥98.0% (by GC)
Thiophene-3-carbaldehyde
Formule:
C₅H₄OS Kookpunt: 198°C Smeltpunt: –30 °C Dichtheid: 1,28 g/cm³ (24 °C) Vlampunt: 75°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00005466 CAS nummer: 498-62-4 EINECS: 207-865-5 |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...