4,6-Dichloropyrimidine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
D2488-25G
D2488-5G
TCIAD2488-25GEA
105
EUR
TCIAD2488-25G
TCIAD2488-5G
4,6-Dichloropyrimidine ≥98.0% (by GC)
4,6-Dichloropyrimidine
Formule:
C₄H₂Cl₂N₂ Kookpunt: 176 °C (1013 hPa) Smeltpunt: 64…66 °C Dichtheid: 1,734 g/cm³ (20 °C) |
MDL Nummer:
MFCD00006109 CAS nummer: 1193-21-1 EINECS: 214-770-2 UN: 3263 ADR: 8,III |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...