4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F0544-1G F0544-5G
TCIAF0544-1GEA 25.3 EUR
TCIAF0544-1G TCIAF0544-5G
4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Formule: C₈H₄F₄O
MW: 192.11 g/mol
Kookpunt: 114
Dichtheid: 1.4
Vlampunt: 57°C
MDL Nummer: MFCD00061313
CAS nummer: 90176-80-0
UN: 1989
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a282deb84b3a36eef0e9e173fc21f75dcfdf7b2&vid=2a282deb84b3a36eef0e9e173fc21f75dcfdf7b2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %