Butyl levulinate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L0138-25ML
L0138-500ML
TCIAL0138-25MLEA
19.8
EUR
TCIAL0138-25ML
TCIAL0138-500ML
Butyl levulinate ≥98.0% (by GC)
Butyl levulinate
Formule:
C₉H₁₆O₃ Kookpunt: 237°C Dichtheid: 0,9975 g/cm³ (13 °C) Vlampunt: 120°C |
MDL Nummer:
MFCD00009449 CAS nummer: 2052-15-5 EINECS: 218-143-4 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...