Ethyl levulinate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L0120-25ML L0120-500ML
TCIAL0120-25MLEA 17.6 EUR
TCIAL0120-25ML TCIAL0120-500ML
Ethyl levulinate ≥98.0% (by GC)
Ethyl levulinate
Formule: C₇H₁₂O₃
Kookpunt: 93 °C (1013 hPa)
Dichtheid: 1,01 g/cm³ (20 °C)
Vlampunt: 100°C
MDL Nummer: MFCD00009209
CAS nummer: 539-88-8
EINECS: 208-728-2
Merck Index: 14,03819
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55dcd17cabb256540674f164a8929df989dc4060&vid=55dcd17cabb256540674f164a8929df989dc4060&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR