5-Bromo-2,4-dichloropyrimidine ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B2823-25G B2823-5G
TCIAB2823-25GEA 223 EUR
TCIAB2823-25G TCIAB2823-5G
5-Bromo-2,4-dichloropyrimidine ≥97.0% (by GC)
5-Bromo-2,4-dichloropyrimidine
Formule: C₄HBrCl₂N₂
MW: 227.87 g/mol
Kookpunt: 288
Smeltpunt: -1
MDL Nummer: MFCD00127818
CAS nummer: 36082-50-5
UN: 2923
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c391db09ff380877552ce52f6e7579b2980f840&vid=3c391db09ff380877552ce52f6e7579b2980f840&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %