5-Chloro-2-methylindole ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1761-1G C1761-5G
TCIAC1761-1GEA 47.3 EUR
TCIAC1761-1G TCIAC1761-5G
5-Chloro-2-methylindole ≥98.0% (by GC, titration analysis)
5-Chloro-2-methylindole
Formule: C₉H₈ClN
Kookpunt: 140°C
Smeltpunt: 129 °C
MDL Nummer: MFCD00005619
CAS nummer: 1075-35-0
EINECS: 214-052-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cfb0f923cd302af0e1eef47d74b255e5194e04f&vid=0cfb0f923cd302af0e1eef47d74b255e5194e04f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C