5-Chlorovaleryl chloride ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C1789-25G C1789-5G
TCIAC1789-25GEA 60.5 EUR
TCIAC1789-25G TCIAC1789-5G
5-Chlorovaleryl chloride ≥98.0% (by GC, titration analysis)
5-Chlorovaleryl chloride
Formule: C₅H₈Cl₂O
Kookpunt: 97 - 99 °C (1 -rr)
Dichtheid: 1,21 g/cm³ (20 °C)
Vlampunt: 90°C
MDL Nummer: MFCD00000758
CAS nummer: 1575-61-7
EINECS: 216-403-1
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e5ce0237f41a86ab23cef4fef6baf7117f57b0b&vid=4e5ce0237f41a86ab23cef4fef6baf7117f57b0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR