Benzofuran ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone
B0060-25G
B0060-5G
TCIAB0060-25GEA
97.9
EUR
TCIAB0060-25G
TCIAB0060-5G
Benzofuran ≥99.0% (by GC)
Benzofuran
Formule:
C₈H₆O Kookpunt: 173 °C (1013 hPa) Smeltpunt: < –18 °C Dichtheid: 1,1 g/cm³ (20 °C) Vlampunt: 56°C |
MDL Nummer:
MFCD00005847 CAS nummer: 271-89-6 EINECS: 205-982-6 UN: 1993 ADR: 3,III Merck Index: 14,01088 |
|
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...