Benzyl bromoacetate ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Benzyl 2-bromoacetate, Bromoacetic benzyl ester
B1674-25G B1674-500G
TCIAB1674-25GEA 19.8 EUR
TCIAB1674-25G TCIAB1674-500G
Benzyl bromoacetate ≥96.0% (by GC)
Benzyl bromoacetate
Formule: C₉H₉BrO₂
MW: 229.07 g/mol
Kookpunt: 170°C
MDL Nummer: MFCD00000190
CAS nummer: 5437-45-6
EINECS: 226-611-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=906686bcc36586a91e64fc52c466dfa698b7a397&vid=906686bcc36586a91e64fc52c466dfa698b7a397&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %