tert-Butyl chloroacetate ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1267-25ML C1267-500ML
TCIAC1267-25MLEA 24.2 EUR
TCIAC1267-25ML TCIAC1267-500ML
tert-Butyl chloroacetate ≥97.0% (by GC)
tert-Butyl chloroacetate
Formule: C₆H₁₁ClO₂
Kookpunt: 60°C
Dichtheid: 1,053 g/cm³ (20 °C)
Vlampunt: 28°C
MDL Nummer: MFCD00000930
CAS nummer: 107-59-5
EINECS: 203-506-1
UN: 3272
ADR: 3,III
Merck Index: 14,01563
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc20e6cd3c9d4c39b6f7258f53745c8a17f2dfa1&vid=dc20e6cd3c9d4c39b6f7258f53745c8a17f2dfa1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %