Ethylchlooracetaat ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C0092-25ML C0092-500ML
TCIAC0092-25MLEA 17.6 EUR
TCIAC0092-25ML TCIAC0092-500ML
Ethylchlooracetaat ≥98.0% (by GC)
Ethylchlooracetaat
Formule: C₄H₇ClO₂
Kookpunt: 143°C
Smeltpunt: –26 °C
Dichtheid: 1,15 g/cm³ (20 °C)
Vlampunt: 57°C
MDL Nummer: MFCD00000932
CAS nummer: 105-39-5
EINECS: 203-294-0
UN: 1181
ADR: 6.1,II
Merck Index: 14,03783
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5ed0c1a69653b3ebec5a5f312d718069243c4af&vid=b5ed0c1a69653b3ebec5a5f312d718069243c4af&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 98.0 %