Oleyl amine >50.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: cis-9-Octadecenylamine, (9Z)-9-Octadecen-1-amine, (Z)-Octadec-9-en-1-amine
O0059-25ML O0059-500ML
TCIAO0059-25MLEA 17.6 EUR
TCIAO0059-25ML TCIAO0059-500ML
Oleyl amine >50.0% (by GC)
Oleyl amine
Formule: C₁₈H₃₇N
Kookpunt: 364 °C (1013 hPa)
Smeltpunt: 21 °C
Dichtheid: 0,813 g/cm³ (20 °C)
Vlampunt: 158°C
MDL Nummer: MFCD00066507
CAS nummer: 112-90-3
EINECS: 204-015-5
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aea92ea7e4ab5fcbff6381e6d8c20b118d771162&vid=aea92ea7e4ab5fcbff6381e6d8c20b118d771162&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 50.0 %