Maleimide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1H-Pyrrole-2,5-dione

M0018-25G M0018-5G
TCIAM0018-25GEA 165 EUR
TCIAM0018-25G TCIAM0018-5G
Maleimide ≥98.0% (by titrimetric analysis)
Maleimide
Formule: C₄H₃NO₂
Kookpunt: 125°C
Smeltpunt: 90…93 °C
Dichtheid: 1,2493 g/cm³ (106 °C)
MDL Nummer: MFCD00005494
CAS nummer: 541-59-3
EINECS: 208-787-4
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d318089a16121dcd366c3a85e93bdbeedfff951b&vid=d318089a16121dcd366c3a85e93bdbeedfff951b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 92.0 to 95.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR