Cyanoacetic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0439-25G C0439-500G
TCIAC0439-25GEA 17.6 EUR
TCIAC0439-25G TCIAC0439-500G
Cyanoacetic acid ≥98.0% (by titrimetric analysis)
Cyanoacetic acid
Formule: C₃H₃NO₂
MW: 85.06 g/mol
Kookpunt: 319
Smeltpunt: 66
MDL Nummer: MFCD00002677
CAS nummer: 372-09-8
EINECS: 206-743-9
UN: 3261
ADR: 8 III
Merck Index: 14,02689
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ef2d6aba01aeba29a29e2fb2de3de743d556e3d&vid=5ef2d6aba01aeba29a29e2fb2de3de743d556e3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 73.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR