Isopropyl cyanoacetate ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Isopropyl 2-cyanoacetate
C1001-25ML C1001-500ML
TCIAC1001-25MLEA 18.7 EUR
TCIAC1001-25ML TCIAC1001-500ML
Isopropyl cyanoacetate ≥97.0% (by GC)
Isopropyl cyanoacetate
Formule: C₆H₉NO₂
MW: 127.14 g/mol
Kookpunt: 206°C
Vlampunt: 99°C
MDL Nummer: MFCD00019842
CAS nummer: 13361-30-3
EINECS: 236-421-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f119e069ce51c02523dc5d2fdfc2ef18ad4b552&vid=6f119e069ce51c02523dc5d2fdfc2ef18ad4b552&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %