Butyl cyanoacetate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Cyanoacetic acid butyl ester
C0440-25G C0440-500G
TCIAC0440-25GEA 19.8 EUR
TCIAC0440-25G TCIAC0440-500G
Butyl cyanoacetate ≥98.0% (by GC)
Butyl cyanoacetate
Formule: C₇H₁₁NO₂
MW: 141.17 g/mol
Kookpunt: 231°C
Smeltpunt: -80
Vlampunt: 112°C
MDL Nummer: MFCD00001941
CAS nummer: 5459-58-5
EINECS: 226-730-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7afc9a7c2af235731c3df203be9c70be1c0cd2e6&vid=7afc9a7c2af235731c3df203be9c70be1c0cd2e6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %