Cyclopentyltrichlorosilane ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T2576-5G
T2576-25G
TCIAT2576-5GEA
27.5
EUR
TCIAT2576-5G
TCIAT2576-25G
Cyclopentyltrichlorosilane ≥95.0% (by GC)
Cyclopentyltrichlorosilane
Formule:
C₅H₉Cl₃Si Kookpunt: 178°C Dichtheid: 1,2756 g/cm³ (20 °C) Vlampunt: 68°C |
MDL Nummer:
MFCD00236508 CAS nummer: 14579-03-4 EINECS: 238-621-6 UN: 2987 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...